Learn More
Cayman Chemical Tauroldn 50mg
SDP
Catalog No. 502973286
Encompass_Preferred
A synthetic taurine analog with broad bactericidal and fungicidal activity; binds to the extracellular wall of bacteria, blocking adherence to epithelial and fibroblast cells; induces autophagy, apoptosis, and necrosis in human cancer cells
Catalog No. 50-297-3286 Supplier Cayman Chemical Supplier No. 1537450 MG
A synthetic taurine analog with broad bactericidal and fungicidal activity; binds to the extracellular wall of bacteria, blocking adherence to epithelial and fibroblast cells; induces autophagy, apoptosis, and necrosis in human cancer cells