Learn More
Cayman Chemical TaurolIthocholIc ACD MaxSpec
SDP
Catalog No. 502992646
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-299-2646 Supplier Cayman Chemical Supplier No. 31604100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications