Learn More
Medchemexpress LLC Taurolithocholic acid | 516-90-5 | 99.7% | 483.70 | 10 MG
SDP
Catalog No. 5000436695
Encompass_Preferred
Taurolithocholic acid is an orally active bile acid and antiviral agent. It functions by upregulating FADS2 through the TGR5-PI3K/AKT-SREBP2 signaling axis, inhibiting SFTSV-induced ferroptosis, viral replication, and viral entry of HBV/HDV. It also reduces the release of IL-1β, lipid ROS, and LDH. This compound serves as an experimental model for hepatocellular cholestasis and offers protection against lethal SFTSV infection in mice, making it suitable for related research.
  • Orally active bile acid and antiviral agent
  • Upregulates FADS2 by activating the TGR5-PI3K/AKT-SREBP2 signaling axis
  • Inhibits SFTSV-induced ferroptosis, viral replication, and viral entry of HBV/HDV
  • Reduces the release of IL-1β, lipid ROS, and LDH
  • Experimental model compound for hepatocellular cholestasis
  • Protects mice from lethal SFTSV infection
Catalog No. 50-004-36695 Supplier Medchemexpress LLC Supplier No. HY11330810MG
Add to Cart
Add to Cart
Taurolithocholic acid is an orally active bile acid and antiviral agent. It functions by upregulating FADS2 through the TGR5-PI3K/AKT-SREBP2 signaling axis, inhibiting SFTSV-induced ferroptosis, viral replication, and viral entry of HBV/HDV. It also reduces the release of IL-1β, lipid ROS, and LDH. This compound serves as an experimental model for hepatocellular cholestasis and offers protection against lethal SFTSV infection in mice, making it suitable for related research.
  • Orally active bile acid and antiviral agent
  • Upregulates FADS2 by activating the TGR5-PI3K/AKT-SREBP2 signaling axis
  • Inhibits SFTSV-induced ferroptosis, viral replication, and viral entry of HBV/HDV
  • Reduces the release of IL-1β, lipid ROS, and LDH
  • Experimental model compound for hepatocellular cholestasis
  • Protects mice from lethal SFTSV infection