Learn More
TARGETMOL CHEMICALS INC TAUROLITHOCHOLIC ACID SO 100MG
SDP
Catalog No. 502579850
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg and bulk. Please contact Fisher for quotes. Taurolithocholic acid sodium salt (Sodium taurolithocholate) is the major human metabolite a bile acid which inhibits radioligand binding to muscarinic M1 but not to M2 or M3 receptors. purity: 99%
Catalog No. 50-257-9850 Supplier TARGETMOL CHEMICALS INC Supplier No. T4727100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg and bulk. Please contact Fisher for quotes. Taurolithocholic acid sodium salt (Sodium taurolithocholate) is the major human metabolite a bile acid which inhibits radioligand binding to muscarinic M1 but not to M2 or M3 receptors. purity: 99%