Learn More
Cayman Chemical TaurolIthocholIc AcIdodIM 50mg
SDP
Catalog No. 502967953
Encompass_Preferred
A taurine-conjugated form of lithocholic acid; increases caspase-3 and -7 activity in NTCP-transfected Hep3B cells at 75 µM; has been used to induce cholestasis in ex vivo and in vivo animal models of hepatocellular cholestasis; serum levels of TLCA increase approximately 5-fold in within two hours during an oral lipid tolerance test in humans
Catalog No. 50-296-7953 Supplier Cayman Chemical Supplier No. 1727550 MG
A taurine-conjugated form of lithocholic acid; increases caspase-3 and -7 activity in NTCP-transfected Hep3B cells at 75 µM; has been used to induce cholestasis in ex vivo and in vivo animal models of hepatocellular cholestasis; serum levels of TLCA increase approximately 5-fold in within two hours during an oral lipid tolerance test in humans