Learn More
Cayman Chemical Taurolithocholic Acid 3-sulfate (sodium salt), Purity: ≥95%, CAS: 64936-83-0, 10 mg
SDP
Catalog No. 502967683
Encompass_Preferred
A metabolite of taurolithocholic acid; has been used to study bile acid transport in cellular models and to induce pancreatitis in mouse models of bile acid infusion pancreatitis
Catalog No. 50-296-7683 Supplier Cayman Chemical Supplier No. 1846810 MG
A metabolite of taurolithocholic acid; has been used to study bile acid transport in cellular models and to induce pancreatitis in mouse models of bile acid infusion pancreatitis