Learn More
Medchemexpress LLC HY-19696A 100mg , Tauroursodeoxycholate (sodium) CAS:35807-85-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19696A100MG
Encompass_Preferred
HY-19696A 100mg Tauroursodeoxycholate (sodium) CAS: 35807-85-3 Tauroursodeoxycholate (Tauroursodeoxycholic acid; TUDCA) sodium is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1047