Learn More
Medchemexpress LLC HY-19696 50mg Medchemexpress, Tauroursodeoxycholate CAS:14605-22-2 Purity:>98%
SDP
Catalog No. 502082163
Encompass_Preferred
Medchemexpress, HY-19696 50mg Tauroursodeoxycholate CAS:14605-22-2 Tauroursodeoxycholate (TUDCA; UR 906; Taurolite) is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2163 Supplier Medchemexpress LLC Supplier No. HY1969650MG
Medchemexpress, HY-19696 50mg Tauroursodeoxycholate CAS:14605-22-2 Tauroursodeoxycholate (TUDCA; UR 906; Taurolite) is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.