Learn More
Medchemexpress LLC Tauroursodeoxycholate-d5 | 1207294-25-4 | 504.73 | 5 MG
SDP
Catalog No. 5000400751
Encompass_Preferred
Tauroursodeoxycholate-d5 is the deuterium labeled Tauroursodeoxycholate, an endoplasmic reticulum (ER) stress inhibitor. It significantly reduces the expression of apoptosis molecules, such as caspase-3 and caspase-12, and also inhibits ERK.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-00751 Supplier Medchemexpress LLC Supplier No. HY19696S5MG
Tauroursodeoxycholate-d5 is the deuterium labeled Tauroursodeoxycholate, an endoplasmic reticulum (ER) stress inhibitor. It significantly reduces the expression of apoptosis molecules, such as caspase-3 and caspase-12, and also inhibits ERK.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS