Learn More
Medchemexpress LLC HY-19696A 500mg Medchemexpress, Tauroursodeoxycholate (Sodium) Sodium tauroursodeoxycholate;Tauroursodeoxycholic acid sodium salt CAS:35807-85-3 Purity:98%
SDP
Catalog No. 501638907
Encompass_Preferred
Medchemexpress,HY-19696A 500mg Tauroursodeoxycholate (Sodium) Sodium tauroursodeoxycholate;Tauroursodeoxycholic acid sodium salt CAS:35807-85-3 Formula:C26H44NNaO6S Tauroursodeoxycholate Sodium is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8907 Supplier Medchemexpress LLC Supplier No. HY19696A500MG
Medchemexpress,HY-19696A 500mg Tauroursodeoxycholate (Sodium) Sodium tauroursodeoxycholate;Tauroursodeoxycholic acid sodium salt CAS:35807-85-3 Formula:C26H44NNaO6S Tauroursodeoxycholate Sodium is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.