Learn More
Cayman Chemical Tauroursodeoxycholic Acid (sodium salt), Purity: ≥95%, CAS: 35807-85-3, 500 mg
SDP
Catalog No. 502985572
Encompass_Preferred
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders
Catalog No. 50-298-5572 Supplier Cayman Chemical Supplier No. 20277500 MG
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders