Learn More
Cayman Chemical TauroursodeoxycholIc ACDo 50mg
SDP
Catalog No. 502895348
Encompass_Preferred
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders
Catalog No. 50-289-5348 Supplier Cayman Chemical Supplier No. 900337950 MG
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders