Learn More
Cayman Chemical Tauroursodeoxycholic Acid (sodium salt hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502893967
Encompass_Preferred
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders
Catalog No. 50-289-3967 Supplier Cayman Chemical Supplier No. 9003379100 MG
A taurine-conjugated form of ursodeoxycholic acid; demonstrates anti-apoptotic activity in rodent models of tauopathy, Huntington's disease, ischemic brain injury, and retinal disorders