Learn More
Cayman Chemical Tauroursodeoxycholic Acid, 14605-22-2, 1 g
SDP
Supplier:  Cayman Chemical 202771
Encompass_Preferred
Molecular Formula C26H45NO6S, Purity ≥95%, Formula Weight 499.7, Synonyms: 3α,7β-dihydroxy-5β-cholanoyl Taurine, TUDCA, UR-906; A secondary bile acid with hepatoprotective properties; inhibits apoptosis, in part by interfering with the mitochondrial pathway of cell death, inhibiting ROS, reducing ER stress, and stabilizing the unfolded protein response.
Catalog No. NC1266953