Learn More
Cayman Chemical Tauro-ω-muricholic Acid (sodium salt), Purity: ≥95%, CAS: 2456348-84-6, 10 mg
SDP
Catalog No. 502939798
Encompass_Preferred
A taurine-conjugated form of ω-muricholic acid; hepatic levels are decreased in mice following a diet supplemented with cholic, deoxycholic, chenodeoxycholic, or ursodeoxycholic acid; hepatic levels are decreased in a high-fat diet-induced mouse model of NAFLD; the predominant rare bile acid found in the serum of infants with early-onset neonatal sepsis
Catalog No. 50-293-9798 Supplier Cayman Chemical Supplier No. 2884210 MG
A taurine-conjugated form of ω-muricholic acid; hepatic levels are decreased in mice following a diet supplemented with cholic, deoxycholic, chenodeoxycholic, or ursodeoxycholic acid; hepatic levels are decreased in a high-fat diet-induced mouse model of NAFLD; the predominant rare bile acid found in the serum of infants with early-onset neonatal sepsis