Learn More
Medchemexpress LLC HY-10980 10mg Medchemexpress, Tavaborole CAS:174671-46-6 Purity:>98%
Supplier: Medchemexpress LLC HY1098010MG
Medchemexpress, HY-10980 10mg Tavaborole CAS:174671-46-6 Tavaborole (AN-2690) is an antifungal agent with activity against Trichophyton species, in a topical solution formulation for the potential treatment of onychomycosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.