Learn More
Medchemexpress LLC Tavaborole | 174671-46-6 | 99.7% | 151.93 | 1 ML
SDP
Catalog No. 5000345017
Encompass_Preferred
Tavaborole is an antifungal agent primarily active against Trichophyton species, developed as a topical solution for the treatment of onychomycosis. It is intended strictly for research purposes and not sold to patients.
  • Antifungal agent with activity against Trichophyton species.
  • Used in a topical solution formulation for the potential treatment of onychomycosis.
  • Demonstrates an 8-fold increase in activity against C. neoformans and A. fumigatus in vitro.
  • Inhibits cells expressing GlLeuRS-D444A.
Catalog No. 50-003-45017 Supplier Medchemexpress LLC Supplier No. HY10980200MG
Tavaborole is an antifungal agent primarily active against Trichophyton species, developed as a topical solution for the treatment of onychomycosis. It is intended strictly for research purposes and not sold to patients.
  • Antifungal agent with activity against Trichophyton species.
  • Used in a topical solution formulation for the potential treatment of onychomycosis.
  • Demonstrates an 8-fold increase in activity against C. neoformans and A. fumigatus in vitro.
  • Inhibits cells expressing GlLeuRS-D444A.