Learn More
Cayman Chemical (±)-Taxifolin, Purity: ≥98%, CAS: 24198-97-8, 100 mg
SDP
Catalog No. 502980126
Encompass_Preferred
A flavonoid with diverse biological activities; scavenges DPPH radicals in cell-free assays; cytotoxic to HCT116 cells (IC50 = 63.42 µM); inhibits HMG-CoA reductase activity and cholesterol synthesis in HepG2 cells at 200 µM; reduces carrageenan-induced paw edema in rats (ED50 = 12.9 mg/kg)
Catalog No. 50-298-0126 Supplier Cayman Chemical Supplier No. 18647100 MG
A flavonoid with diverse biological activities; scavenges DPPH radicals in cell-free assays; cytotoxic to HCT116 cells (IC50 = 63.42 µM); inhibits HMG-CoA reductase activity and cholesterol synthesis in HepG2 cells at 200 µM; reduces carrageenan-induced paw edema in rats (ED50 = 12.9 mg/kg)