Learn More
Cayman Chemical (+)-Taxifolin, Purity: ≥95%, CAS: 480-18-2, 100 mg
SDP
Catalog No. 502987217
Encompass_Preferred
A flavonol with antioxidant activity; increases cell viability of FeCl2 and H2O2-treated bmMSCs (1-100 µg/ml); scavenges PTIO radicals in a pH-dependent manner (IC50s = 2.6-0.4 mM for pH 5-9, respectively); protects against H2O2- and xanthine/xanthine oxidase-induced oxidative injury in primary cultured rat cortical cells (IC50 = 7.8 µg/ml); inhibits lipid peroxidation in rat brain homogenates (IC50 = 1.02 µg/ml)
Catalog No. 50-298-7217 Supplier Cayman Chemical Supplier No. 23234100 MG
A flavonol with antioxidant activity; increases cell viability of FeCl2 and H2O2-treated bmMSCs (1-100 µg/ml); scavenges PTIO radicals in a pH-dependent manner (IC50s = 2.6-0.4 mM for pH 5-9, respectively); protects against H2O2- and xanthine/xanthine oxidase-induced oxidative injury in primary cultured rat cortical cells (IC50 = 7.8 µg/ml); inhibits lipid peroxidation in rat brain homogenates (IC50 = 1.02 µg/ml)