Learn More
Apexbio Technology LLC Taxifolin 480-18-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063493
Encompass_Preferred
Taxifolin (CAS number 480-18-2) a bioactive flavonoid isolated primarily from coniferous species of the Pinaceae family exhibits notable antioxidant anti-inflammatory and chemopreventive properties It functions primarily through free radical scavenging and modulation of oxidative stress pathways reducing cellular damage associated with reactive oxygen species (ROS) Taxifolin further influences various signaling cascades involved in inflammation and apoptosis regulation Due to these pharmacological attributes it is widely utilized in biomedical research exploring oxidative stress-related diseases inflammatory conditions and potential cancer therapeutic avenues For optimal stability maintain storage conditions sealed dry and cool
Catalog No. 50-000-63493 Supplier Apexbio Technology LLC Supplier No. N18425.1
Taxifolin (CAS number 480-18-2) a bioactive flavonoid isolated primarily from coniferous species of the Pinaceae family exhibits notable antioxidant anti-inflammatory and chemopreventive properties It functions primarily through free radical scavenging and modulation of oxidative stress pathways reducing cellular damage associated with reactive oxygen species (ROS) Taxifolin further influences various signaling cascades involved in inflammation and apoptosis regulation Due to these pharmacological attributes it is widely utilized in biomedical research exploring oxidative stress-related diseases inflammatory conditions and potential cancer therapeutic avenues For optimal stability maintain storage conditions sealed dry and cool