Learn More
Selleck Chemical LLC Taxifolin (Dihydroquercetin) S2366-50mg
SDP
Supplier:  Selleck Chemical LLC S236650MG
Encompass_Preferred
Taxifolin type I inhibitor for VEGFR-2 kinase is a flavonoid in many plants such as Taxus chinensis Siberian larch Cedrus deodara and so on
Catalog No. 50-311-2345