Learn More
Enzo Life Sciences Taxifolin (50mg). CAS: 17654-26-1
SDP
Supplier:  Enzo Life Sciences ALX385006M050
Encompass_Preferred
Antioxidant flavonoid. Alternative name: (+/-)-Dihydroquercetin, (+/-)-3,3',4',5,7-Pentahydroxyflavanone, Taxifoliol, Distylin, Catechin hydrate. Purity: ≥95% (HPLC). Appearance: White to slightly brown powder. Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9039