Learn More
Selleck Chemical LLC Taxifolin 7-rhamnoside S3295-1mg
SDP
Catalog No. 503111344
Encompass_Preferred
Taxifolin 7-rhamnoside (TR Taxifolin 7-O-rhamnoside Taxifolin 7-O- -L-rhamnoside) a flavonoid isolated from Hypericum japonicum shows antibacterial activity against clinical isolates of methicillin-resistant Staphylococcus aureus (MRSA)
Catalog No. 50-311-1344 Supplier Selleck Chemical LLC Supplier No. S32951MG
Taxifolin 7-rhamnoside (TR Taxifolin 7-O-rhamnoside Taxifolin 7-O- -L-rhamnoside) a flavonoid isolated from Hypericum japonicum shows antibacterial activity against clinical isolates of methicillin-resistant Staphylococcus aureus (MRSA)