Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taxifolin, ≥85% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014264 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4264 25 mg
11-101-4263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4264 Supplier MilliporeSigma Supelco Supplier No. 7866625MGF
Add to Cart
Add to Cart

Taxifolin is a plant flavonoid, which may find applications in the treatment of coronary heart disease, angina pectoris, cerebral thrombosis, cerebral infarction and sequelae.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 480-18-2
Physical Form Neat
Linear Formula C15H12O7
MDL Number MFCD00006845
Synonym (2 R,3 R)-3,3 ′,4 ′,5,7-Pentahydroxyflavanone; (2 R,3 R)-Dihydroquercetin
Type Analytical Standard
Quantity 25 mg
Formula Weight 304.25
Percent Purity ≥85% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.