Learn More
Selleck Chemical LLC Taxifolin (Dihydroquercetin) 200mg 480-18-2
SDP
Supplier:  Selleck Chemical LLC S2366200MG
Encompass_Preferred
Taxifolin, type I inhibitor for VEGFR-2 kinase, is a flavonoid in many plants such as Taxus chinensis, Siberian larch, Cedrus deodara and so on. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-550-9