Learn More
Fine Science Tools Taylor Forceps, Stainless Steel, Straight, 17 cm
SDP
Catalog No. NC9031464
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.05mm/ Alloy / Material: Stainless Steel/ Length: 17cm
Catalog No. NC9031464 Supplier Fine Science Tools Supplier No. 1101017
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.05mm/ Alloy / Material: Stainless Steel/ Length: 17cm