Learn More
Fine Science Tools Taylor Forceps, Stainless Steel, Curved, 17 cm
SDP
Catalog No. NC0357053
Encompass
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 1.5 x 1mm/ Alloy / Material: Stainless Steel/ Length: 17cm
Catalog No. NC0357053 Supplier Fine Science Tools Supplier No. 1101717
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 1.5 x 1mm/ Alloy / Material: Stainless Steel/ Length: 17cm