Learn More
Medchemexpress LLC HY-101810 1mg Medchemexpress, Tazanolast CAS:82989-25-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1018101MG
Encompass_Preferred
Medchemexpress, HY-101810 1mg Tazanolast CAS:82989-25-1 Tazanolast is a selective mast-cell-stabilizing drug, on ozone-induced airway hyperresponsiveness in guinea pigs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7876
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.