Learn More
Medchemexpress LLC HY-15388 10mg Medchemexpress, Tazarotene CAS:118292-40-3 Purity:>98%
SDP
Catalog No. 502081713
Encompass_Preferred
Medchemexpress, HY-15388 10mg Tazarotene CAS:118292-40-3 Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1713 Supplier Medchemexpress LLC Supplier No. HY1538810MG
Medchemexpress, HY-15388 10mg Tazarotene CAS:118292-40-3 Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.