Learn More
Medchemexpress LLC Tazarotene | 118292-40-3 | MFCD00867628 | C21H21NO2S | 1 ML
SDP
Catalog No. 5000376424
Encompass_Preferred
Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist used for treating plaque psoriasis and acne vulgaris. It is also a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Selective retinoic acid receptor (RAR) agonist
  • Used for treating plaque psoriasis
  • Used for treating acne vulgaris
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
Catalog No. 50-003-76424 Supplier Medchemexpress LLC Supplier No. HY1538810MM/1ML
Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist used for treating plaque psoriasis and acne vulgaris. It is also a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Selective retinoic acid receptor (RAR) agonist
  • Used for treating plaque psoriasis
  • Used for treating acne vulgaris
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions