Learn More
Medchemexpress LLC Tazarotene (AGN 190168) | 118292-40-3 | C21H21NO2S | 5 MG
SDP
Catalog No. 5000376315
Encompass_Preferred
Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist used for treating plaque psoriasis and acne vulgaris. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used for monotherapy or adjuvant therapy for psoriasis and acne
  • Upregulates tumor suppressor tazarotene induced gene 3
  • Suppresses gene expression of MRP-8 and SKALP in human epidermal cell cultures
  • Delivered into the skin via topical gel application
  • Metabolized to active tazarotenic acid with short systemic residence time
  • Blocks induction of ornithine decarboxylase activity
Catalog No. 50-003-76315 Supplier Medchemexpress LLC Supplier No. HY153885MG
Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist used for treating plaque psoriasis and acne vulgaris. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used for monotherapy or adjuvant therapy for psoriasis and acne
  • Upregulates tumor suppressor tazarotene induced gene 3
  • Suppresses gene expression of MRP-8 and SKALP in human epidermal cell cultures
  • Delivered into the skin via topical gel application
  • Metabolized to active tazarotenic acid with short systemic residence time
  • Blocks induction of ornithine decarboxylase activity