Learn More
Medchemexpress LLC Tazarotene (Standard) | 118292-40-3 | C21H21NO2S | 5 MG
SDP
Catalog No. 5000376699
Encompass_Preferred
Tazarotene (Standard) is the analytical standard of Tazarotene, intended for research and analytical applications. It acts as a selective retinoic acid receptor (RAR) agonist, used in the treatment of plaque psoriasis and acne vulgaris. As a click chemistry reagent, it contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Analytical standard for research and analytical applications
  • Selective retinoic acid receptor (RAR) agonist
  • Treats plaque psoriasis and acne vulgaris
  • Click chemistry reagent with alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
Catalog No. 50-003-76699 Supplier Medchemexpress LLC Supplier No. HY15388R5MG
Tazarotene (Standard) is the analytical standard of Tazarotene, intended for research and analytical applications. It acts as a selective retinoic acid receptor (RAR) agonist, used in the treatment of plaque psoriasis and acne vulgaris. As a click chemistry reagent, it contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Analytical standard for research and analytical applications
  • Selective retinoic acid receptor (RAR) agonist
  • Treats plaque psoriasis and acne vulgaris
  • Click chemistry reagent with alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions