Learn More
Medchemexpress LLC Tazarotene (standard) | 118292-40-3 | C21H21NO2S | 50 MG
SDP
Catalog No. 5000376516
Encompass_Preferred
Tazarotene (Standard) is an analytical standard intended for research and analytical applications. It acts as a selective retinoic acid receptor (RAR) agonist, used in the treatment of plaque psoriasis and acne vulgaris. It is also a click chemistry reagent, containing an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have azide groups.
  • Analytical standard for research and analytical applications
  • Selective retinoic acid receptor (RAR) agonist
  • Used in the treatment of plaque psoriasis and acne vulgaris
  • Click chemistry reagent containing an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-003-76516 Supplier Medchemexpress LLC Supplier No. HY15388R50MG
Tazarotene (Standard) is an analytical standard intended for research and analytical applications. It acts as a selective retinoic acid receptor (RAR) agonist, used in the treatment of plaque psoriasis and acne vulgaris. It is also a click chemistry reagent, containing an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have azide groups.
  • Analytical standard for research and analytical applications
  • Selective retinoic acid receptor (RAR) agonist
  • Used in the treatment of plaque psoriasis and acne vulgaris
  • Click chemistry reagent containing an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS