Learn More
Selleck Chemical LLC Tazarotene 50mg 118292-40-3 AGN190168
SDP
Supplier:  Selleck Chemical LLC S156950MG
Encompass_Preferred
Tazarotene (AGN190168) is a retinoid prodrug of tazarotenic acid, which is a RAR agonist, used to treat psoriasis, acne, and sun damaged skin. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-771-2