Learn More
Cayman Chemical Tazarotenic Acid, Purity: ≥98%, CAS: 118292-41-4, 50 mg
SDP
Catalog No. 502981485
Encompass_Preferred
An RAR agonist and an active metabolite of tazarotene; induces reporter gene expression in DK7-NR cells expressing human RARα, RARβ, or RARγ (EC50s = 11, 2.5, and 11 nM, respectively); reduces ear edema in a PMA-induced mouse model of epidermal inflammation when applied topically at 0.1% w/w
Catalog No. 50-298-1485 Supplier Cayman Chemical Supplier No. 2136750 MG
An RAR agonist and an active metabolite of tazarotene; induces reporter gene expression in DK7-NR cells expressing human RARα, RARβ, or RARγ (EC50s = 11, 2.5, and 11 nM, respectively); reduces ear edema in a PMA-induced mouse model of epidermal inflammation when applied topically at 0.1% w/w