Learn More
TARGETMOL CHEMICALS INC TAZEMETOSTAT HYDROBROMID 100MG
SDP
Catalog No. 502581132
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tazemetostat hydrobromide (E-7438 hydrobromide) is a potent and selective EZH2 inhibitor. Tazemetostat hydrobromide inhibits the activity of human polycomb repressive complex 2 (PRC2)-containing wild-type EZH2 (Ki 2.5 nM). Tazemetostat hydrobromide also inhibits EZH1 (IC50 392 nM). purity: 99%
Catalog No. 50-258-1132 Supplier TARGETMOL CHEMICALS INC Supplier No. T17002100MG
Also available in 2 mg 5 mg 10 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tazemetostat hydrobromide (E-7438 hydrobromide) is a potent and selective EZH2 inhibitor. Tazemetostat hydrobromide inhibits the activity of human polycomb repressive complex 2 (PRC2)-containing wild-type EZH2 (Ki 2.5 nM). Tazemetostat hydrobromide also inhibits EZH1 (IC50 392 nM). purity: 99%