Learn More
TARGETMOL CHEMICALS INC Tazobactam 1G
SDP
Catalog No. 502875940
Encompass_Preferred
Also available in 1 mL, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tazobactam (YTR-830H) is an antibacterial penicillin derivative which inhibits the action of bacterial beta-lactamases. Purity 100%
Catalog No. 50-287-5940 Supplier TARGETMOL CHEMICALS INC Supplier No. T12621G
Also available in 1 mL, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tazobactam (YTR-830H) is an antibacterial penicillin derivative which inhibits the action of bacterial beta-lactamases. Purity 100%