Learn More
Selleck Chemical LLC Tazobactam S3077-100mg
SDP
Supplier:  Selleck Chemical LLC S3077100MG
Encompass_Preferred
Tazobactam (Tazobactam acid Tazobactamum) is a -lactamases Inhibitor with antibacterial activity It also inhibits the class D oxacillinase OXA-2 It is used in combination with piperacillin and other -lactam antibiotics to broaden their spectrum and enhance their effect
Catalog No. 50-310-8889