Learn More
Selleck Chemical LLC Tazobactam S3077-500mg
SDP
Catalog No. 503110101
Encompass_Preferred
Tazobactam (Tazobactam acid Tazobactamum) is a -lactamases Inhibitor with antibacterial activity It also inhibits the class D oxacillinase OXA-2 It is used in combination with piperacillin and other -lactam antibiotics to broaden their spectrum and enhance their effect
Catalog No. 50-311-0101 Supplier Selleck Chemical LLC Supplier No. S3077500MG
Tazobactam (Tazobactam acid Tazobactamum) is a -lactamases Inhibitor with antibacterial activity It also inhibits the class D oxacillinase OXA-2 It is used in combination with piperacillin and other -lactam antibiotics to broaden their spectrum and enhance their effect