Learn More
Medchemexpress LLC Tazobactam | 89786-04-9 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB14181G
Encompass_Preferred
Tazobactam (CL-298741) is a potent β-lactamase inhibitor and penicillin antibiotic with antibacterial activity, suitable for pneumonia research.
  • Shows bactericidal activity.
  • Inhibits bacterial growth in a dose-dependent manner.
  • Inhibits various types of β-lactamase activity.
  • Decreases the number of bacteria in animal models.
  • Allows survival at low doses in animal models.
Catalog No. 50-004-13406