Learn More
Medchemexpress LLC Tazobactam | 89786-04-9 | 5 G
SDP
Catalog No. 5000412704
Encompass_Preferred
Tazobactam (CL-298741) is a potent β-lactamases inhibitor and a penicillin antibiotic. It exhibits antibacterial activity and can be used for pneumonia research. This product is intended for research and analytical applications.
  • Potent β-lactamases inhibitor
  • Penicillin antibiotic
  • Exhibits antibacterial activity
  • Can be used for pneumonia research
  • Intended for research and analytical applications
Catalog No. 50-004-12704 Supplier Medchemexpress LLC Supplier No. HYB14185G
Tazobactam (CL-298741) is a potent β-lactamases inhibitor and a penicillin antibiotic. It exhibits antibacterial activity and can be used for pneumonia research. This product is intended for research and analytical applications.
  • Potent β-lactamases inhibitor
  • Penicillin antibiotic
  • Exhibits antibacterial activity
  • Can be used for pneumonia research
  • Intended for research and analytical applications