Learn More
Medchemexpress LLC Tazobactam | 89786-04-9 | 99.9% | 1 ML
SDP
Catalog No. 5000412703
Encompass_Preferred
Tazobactam is a potent β-lactamases inhibitor and penicillin antibiotic. It exhibits antibacterial activity and can be used for pneumonia research.
  • Potent β-lactamases inhibitor
  • Penicillin antibiotic
  • Antibacterial activity
  • Can be used for pneumonia research
Catalog No. 50-004-12703 Supplier Medchemexpress LLC Supplier No. HYB141810MM/1ML
Tazobactam is a potent β-lactamases inhibitor and penicillin antibiotic. It exhibits antibacterial activity and can be used for pneumonia research.
  • Potent β-lactamases inhibitor
  • Penicillin antibiotic
  • Antibacterial activity
  • Can be used for pneumonia research