Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tazobactam, G-Biosciences
SCI_ED

Catalog No. S139722
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139722 5 g
S139768 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139722 Supplier G-Biosciences Supplier No. RC1110
Add to Cart
Add to Cart

Tazobactam is a Beta-lactamase inhibitor useful in combination with Beta-lactam antibiotics to increase their potency.

It irreversibly inactivates Beta-lactamase enzymes, preventing them from degrading Beta-lactam antibiotics. Tazobactam has been used in combination with piperacillin and a variety of other antibiotics.

Chemical Identifiers

Molecular Weight (g/mol) 300.29 g/mol

Specifications

Boiling Point 77°C
Quantity 5 g
Percent Purity 95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.