Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tazobactam Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015761 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5761 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5761 Supplier MilliporeSigma Supelco Supplier No. PHR16861G
Add to Cart
Add to Cart

Tazobactam is a penicillanic acid sulfone used in combination with β-lactamase antibiotic as antibacterial to reduce the development of drug-resistant bacteria.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 89786-04-9
Linear Formula C10H12N4O5 S
Synonym Tazobactam; [2 S-(2 alpha,3 beta,5 alpha)]-3-Methyl-7-oxo-3-(1 H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0 ]heptan-2-carbonsäure-4,4-dioxid
Type Certified Reference Material
Quantity 1 g
Formula Weight 300.29
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.