Learn More
Selleck Chemical LLC Tazobactam Sodium-E0167-25MG
SDP
Catalog No. 503138512
Encompass_Preferred
Tazobactam (Tazobactam acid Tazobactamum) Sodium is a beta-lactamase inhibitor that prevents the breakdown of other antibiotics by beta-lactamase enzyme producing organisms
Catalog No. 50-313-8512 Supplier Selleck Chemical LLC Supplier No. E016725MG
Tazobactam (Tazobactam acid Tazobactamum) Sodium is a beta-lactamase inhibitor that prevents the breakdown of other antibiotics by beta-lactamase enzyme producing organisms