Learn More
Selleck Chemical LLC Tazobactam Sodium-E0167-5MG
SDP
Catalog No. 503136854
Encompass_Preferred
Tazobactam (Tazobactam acid Tazobactamum) Sodium is a beta-lactamase inhibitor that prevents the breakdown of other antibiotics by beta-lactamase enzyme producing organisms
Catalog No. 50-313-6854 Supplier Selleck Chemical LLC Supplier No. E01675MG
Tazobactam (Tazobactam acid Tazobactamum) Sodium is a beta-lactamase inhibitor that prevents the breakdown of other antibiotics by beta-lactamase enzyme producing organisms