Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tazobactam Sodium, G-Biosciences
SCI_ED

Catalog No. S157234
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157234 500 mg
S139741 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157234 Supplier G-Biosciences Supplier No. RC1112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tazobactam is a Beta-lactamase inhibitor derived from penicillin. It is used along with Beta-lactam antibiotics, to prevent their degradation by Beta-lactamase enzymes.

Tazobactam is often used with piperacillin to increase its potency. Additionally, tazobactam is used in studies regarding Beta-lactamase and penicillinase inhibition.

Chemical Identifiers

Molecular Weight (g/mol) 322.27 g/mol

Specifications

Boiling Point 77°C
Quantity 500 mg
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.