Learn More
Apexbio Technology LLC Tazobactam (sodium salt) 89785-84-2 1g
SDP
Catalog No. 5000091623
Encompass_Preferred
Tazobactam sodium salt (CAS 89785-84-2) is a small-molecule inhibitor targeting -lactamases It is designed to irreversibly bind to -lactamases thereby preventing the enzyme-mediated hydrolysis of -lactam antibiotics Tazobactam sodium salt exerts its biological activity primarily through -lactamase inhibition In microbiological assays tazobactam demonstrates intrinsic antibacterial activity against several Acinetobacter strains typically with MIC values of 8 mg/L (range 2 32 mg/L) Based on these pharmacological properties tazobactam sodium salt holds research potential in microbiological assessments and efficacy evaluations of -lactam antibiotic combinations
Catalog No. 50-000-91623 Supplier Apexbio Technology LLC Supplier No. C42451000
Tazobactam sodium salt (CAS 89785-84-2) is a small-molecule inhibitor targeting -lactamases It is designed to irreversibly bind to -lactamases thereby preventing the enzyme-mediated hydrolysis of -lactam antibiotics Tazobactam sodium salt exerts its biological activity primarily through -lactamase inhibition In microbiological assays tazobactam demonstrates intrinsic antibacterial activity against several Acinetobacter strains typically with MIC values of 8 mg/L (range 2 32 mg/L) Based on these pharmacological properties tazobactam sodium salt holds research potential in microbiological assessments and efficacy evaluations of -lactam antibiotic combinations