Learn More
Alkali Scientific TRAzol, RNA Purification Reagent Kit, 100ml, Store at 2-8°C, protect from light
SDP
Catalog No. 502336208
Encompass_Preferred
TRAzol, RNA Purification Reagent Kit, 100ml TRAzol Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from cell and tissue samples of human, animal, plant, yeast, or bacterial origin, within one hour. Key features of TRAzol Reagent include: • Permits the isolation of RNA, DNA, and protein from the same sample • Offers superior lysis capability, even with difficult sample types • Optimized formulations and protocols for tissues, cells, serum, virus, and bacteria Reliably purify RNA from multiple sample volumes and sources TRAzol Reagent performs well with small quantities of tissue (50–100 mg) and cells (5 × 106), as well as with large quantities of tissue (≥1 g) and cells (>107), and comes with prototcols for purific
Catalog No. 50-233-6208 Supplier Alkali Scientific Supplier No. TRZ100
TRAzol, RNA Purification Reagent Kit, 100ml TRAzol Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from cell and tissue samples of human, animal, plant, yeast, or bacterial origin, within one hour. Key features of TRAzol Reagent include: • Permits the isolation of RNA, DNA, and protein from the same sample • Offers superior lysis capability, even with difficult sample types • Optimized formulations and protocols for tissues, cells, serum, virus, and bacteria Reliably purify RNA from multiple sample volumes and sources TRAzol Reagent performs well with small quantities of tissue (50–100 mg) and cells (5 × 106), as well as with large quantities of tissue (≥1 g) and cells (>107), and comes with prototcols for purific