Learn More
MTC Bio Conical Centrifuge Tube 50mL in Eco-Plastic Racks
Catalog No. NC2853783
Encompass_Preferred
Conical centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Eco-Plastic racks are a recyclable alternative to standard foam racks when it's time for disposal, or can be broken down into a compact, flat package that is easy to store between uses. • Sterile, RNase, DNase, pyrogen and cytotoxin free• Compatible with standard rotors• Frosted marking surface
Catalog No. NC2853783 Supplier MTC Bio Supplier No. C2608
Conical centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Eco-Plastic racks are a recyclable alternative to standard foam racks when it's time for disposal, or can be broken down into a compact, flat package that is easy to store between uses. • Sterile, RNase, DNase, pyrogen and cytotoxin free• Compatible with standard rotors• Frosted marking surface